Anti-CDKN2A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301407EA 498 EUR
ORIGTA301407
Anti-CDKN2A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR