Anti-HSD3B1 Mouse Monoclonal Antibody [clone: FDO66Q]
Lieferant: OriGene
ORIGTA301417EA 658 EUR
ORIGTA301417
Anti-HSD3B1 Mouse Monoclonal Antibody [clone: FDO66Q]
Antikörper
  • Recommended Dilutions: Western Blot: 1: 1000
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen