Anti-PTGDS Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301420EA 658 EUR
ORIGTA301420
Anti-PTGDS Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:2 ?g/ml

Type: Primary
Antigen: PTGDS (prostaglandin D2 synthase 21kDa (brain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR