Anti-PLIN2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301422EA 599 EUR
ORIGTA301422
Anti-PLIN2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: PLIN2 (perilipin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR