Anti-APBB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301423EA 498 EUR
ORIGTA301423
Anti-APBB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR