Anti-BIRC5 Mouse Monoclonal Antibody [clone: 60.11]

Lieferant: OriGene
ORIGTA301425EA 624 EUR
ORIGTA301425
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 60.11]
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR