Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]

Lieferant: OriGene
ORIGTA301427EA 526 EUR
ORIGTA301427
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR