Anti-BIRC5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301429EA 658 EUR
ORIGTA301429
Anti-BIRC5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:3 ?g/ml

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR