Anti-ARNTL Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301430EA 711 EUR
ORIGTA301430
Anti-ARNTL Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 100-1:200

Type: Primary
Antigen: ARNTL (aryl hydrocarbon receptor nuclear translocator-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR