Anti-CA9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301431EA 747 EUR
ORIGTA301431
Anti-CA9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000; Immunochemistry: 1: 1000; Immunofluorescence: 1: 1000

Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR