Anti-HSPB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301439EA 526 EUR
ORIGTA301439
Anti-HSPB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR