Anti-APEX1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301441EA 599 EUR
ORIGTA301441
Anti-APEX1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR