Anti-COX4I1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301446EA 711 EUR
ORIGTA301446
Anti-COX4I1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: COX4I1 (cytochrome c oxidase subunit IV isoform 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR