Anti-LOX Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301450EA 526 EUR
ORIGTA301450
Anti-LOX Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR