Anti-PIK3C3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301456EA 526 EUR
ORIGTA301456
Anti-PIK3C3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: PIK3C3 (phosphatidylinositol 3-kinase, catalytic subunit type 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR