Anti-POU5F1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301457EA 570 EUR
ORIGTA301457
Anti-POU5F1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2?g/ml

Type: Primary
Antigen: POU5F1 (POU class 5 homeobox 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR