Anti-ABCE1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301458EA 526 EUR
ORIGTA301458
Anti-ABCE1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:250-1: 1000

Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR