Anti-S100A9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301459EA 747 EUR
ORIGTA301459
Anti-S100A9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: S100A9 (S100 calcium binding protein A9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR