Anti-STIM1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301466EA 624 EUR
ORIGTA301466
Anti-STIM1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: STIM1 (stromal interaction molecule 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR