Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]

Lieferant: OriGene
ORIGTA301468EA 624 EUR
ORIGTA301468
Anti-THOP1 Mouse Monoclonal Antibody [clone: 4D6]
Antikörper
Recommended Dilutions: Western Blot: 1:750-1:3000

Type: Primary
Antigen: THOP1 (thimet oligopeptidase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR