Anti-BECN1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301470EA 599 EUR
ORIGTA301470
Anti-BECN1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:2000-1:5000

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR