Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]

Lieferant: OriGene
ORIGTA301475EA 526 EUR
ORIGTA301475
Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: AIP (aryl hydrocarbon receptor interacting protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR