Anti-UCHL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301476EA 658 EUR
ORIGTA301476
Anti-UCHL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR