Anti-KIF11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301478EA 711 EUR
ORIGTA301478
Anti-KIF11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: KIF11 (kinesin family member 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR