Anti-PODXL Mouse Monoclonal Antibody [clone: TRA-1-81]
Lieferant: OriGene
ORIGTA301485EA 624 EUR
ORIGTA301485
Anti-PODXL Mouse Monoclonal Antibody [clone: TRA-1-81]
Antikörper
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: PODXL (podocalyxin-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human