Anti-XRCC3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301486EA 526 EUR
ORIGTA301486
Anti-XRCC3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:5000

Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR