Anti-SCARB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301489EA 599 EUR
ORIGTA301489
Anti-SCARB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000-1:5000

Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR