Anti-RAD50 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301492EA 498 EUR
ORIGTA301492
Anti-RAD50 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000-1: 10000

Type: Primary
Antigen: RAD50 (RAD50 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR