Anti-MAT2A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301494EA 526 EUR
ORIGTA301494
Anti-MAT2A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: MAT2A (methionine adenosyltransferase II, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR