Anti-LIPG Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301495EA 498 EUR
ORIGTA301495
Anti-LIPG Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: LIPG (lipase, endothelial)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR