Anti-RAD21 Mouse Monoclonal Antibody [clone: CM110-2C10]

Lieferant: OriGene
ORIGTA301500EA 498 EUR
ORIGTA301500
Anti-RAD21 Mouse Monoclonal Antibody [clone: CM110-2C10]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR