Anti-CELF1 Mouse Monoclonal Antibody [clone: 3B1]

Lieferant: OriGene
ORIGTA301501EA 599 EUR
ORIGTA301501
Anti-CELF1 Mouse Monoclonal Antibody [clone: 3B1]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CELF1 (CUGBP, Elav-like family member 1)
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR