Anti-OS9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301503EA 624 EUR
ORIGTA301503
Anti-OS9 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.2-1.0 μg/ml

Type: Primary
Antigen: OS9 (osteosarcoma amplified 9, endoplasmic reticulum lectin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR