Anti-ABCF2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301507EA 498 EUR
ORIGTA301507
Anti-ABCF2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: ABCF2 (ATP-binding cassette, sub-family F (GCN20), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR