Anti-TARDBP Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301509EA 498 EUR
ORIGTA301509
Anti-TARDBP Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR