Anti-DNM1L Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301511EA 599 EUR
ORIGTA301511
Anti-DNM1L Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: DNM1L (dynamin 1-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR