Anti-FOXP3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301514EA 570 EUR
ORIGTA301514
Anti-FOXP3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000-1:5000

Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR