Anti-BBC3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301519EA 624 EUR
ORIGTA301519
Anti-BBC3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: BBC3 (BCL2 binding component 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR