Anti-POT1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301520EA 658 EUR
ORIGTA301520
Anti-POT1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: POT1 (protection of telomeres 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR