Anti-ABHD5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301522EA 498 EUR
ORIGTA301522
Anti-ABHD5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ABHD5 (abhydrolase domain containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR