Anti-SIRT6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301523EA 526 EUR
ORIGTA301523
Anti-SIRT6 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5-2 μg/ml

Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR