Anti-IFT57 Mouse Monoclonal Antibody [clone1E1]

Lieferant: OriGene
ORIGTA301535EA 498 EUR
ORIGTA301535
Anti-IFT57 Mouse Monoclonal Antibody [clone1E1]
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: IFT57 (intraflagellar transport 57 homolog (Chlamydomonas))
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR