Anti-MAP1LC3B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301541EA 526 EUR
ORIGTA301541
Anti-MAP1LC3B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: MAP1LC3B (microtubule-associated protein 1 light chain 3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR