Anti-DERL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301554EA 498 EUR
ORIGTA301554
Anti-DERL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000-1:5000

Type: Primary
Antigen: DERL1 (derlin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR