Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]

Lieferant: OriGene
ORIGTA301555EA 570 EUR
ORIGTA301555
Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: WT1 (Wilms tumor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR