Anti-FBXO44 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301558EA 429 EUR
ORIGTA301558
Anti-FBXO44 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: FBXO44 (F-box protein 44)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR