Anti-APOA5 Mouse Monoclonal Antibody [clone: 1G5G9]

Lieferant: OriGene
ORIGTA301561EA 498 EUR
ORIGTA301561
Anti-APOA5 Mouse Monoclonal Antibody [clone: 1G5G9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR