Anti-CREB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301564EA 701 EUR
ORIGTA301564
Anti-CREB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation:5 ?g/ml

Type: Primary
Antigen: CREB1 (cAMP responsive element binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR