Anti-TERT Mouse Monoclonal Antibody [clone: 2C4]

Lieferant: OriGene
ORIGTA301588EA 598 EUR
ORIGTA301588
Anti-TERT Mouse Monoclonal Antibody [clone: 2C4]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR