Anti-LPIN1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301605EA 566 EUR
ORIGTA301605
Anti-LPIN1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: LPIN1 (lipin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR